Assay Details
| Gene Symbol: sri-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073239.3 | NP_505640.2 | 1 - 2 | 169 | 167 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sri-18| Gene Symbol: sri-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073239.3 | NP_505640.2 | 1 - 2 | 169 | 167 | |
Set Membership: |
|