Assay Details
| Gene Symbol: che-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269275.3 | NP_001256204.1 | 17 - 18 | 2820 | 116 | |
| NM_001269276.3 | NP_001256205.1 | 8 - 9 | 1205 | 116 | ||
| NM_001269277.3 | NP_001256206.1 | 6 - 7 | 748 | 116 | ||
| NM_001307803.4 | NP_001294732.1 | 6 - 7 | 788 | 116 | ||
More Information
Set Membership: |
|

che-12
Genomic Map