Assay Details
| Gene Symbol: ttll-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073262.7 | NP_505663.3 | 10 - 11 | 1260 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ttll-15| Gene Symbol: ttll-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073262.7 | NP_505663.3 | 10 - 11 | 1260 | 99 | |
Set Membership: |
|