Assay Details
| Gene Symbol: sri-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001355085.1 | NP_001342025.1 | 3 - 4 | 513 | 89 | |
| NM_001373097.1 | NP_001359573.1 | 3 - 4 | 540 | 89 | ||
More Information
Set Membership: |
|

sri-20
Genomic Map