Assay Details
| Gene Symbol: col-151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073269.2 | NP_505670.1 | 1 - 2 | 153 | 149 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

col-151| Gene Symbol: col-151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073269.2 | NP_505670.1 | 1 - 2 | 153 | 149 | |
Set Membership: |
|