Assay Details
| Gene Symbol: set-22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380744.2 | NP_001366641.1 | 1 - 2 | 131 | 124 | |
| Gene Symbol: pap-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383385.1 | 1 - 2 | 175 | 124 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383385.1 transcripts.
Set Membership: |
|

set-22
Genomic Map