Assay Details
| Gene Symbol: cat-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073309.6 | NP_505710.1 | 2 - 3 | 290 | 74 | |
| GenBank mRNA | KP290890.1 | - | 2 - 3 | 316 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cat-4| Gene Symbol: cat-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073309.6 | NP_505710.1 | 2 - 3 | 290 | 74 | |
| GenBank mRNA | KP290890.1 | - | 2 - 3 | 316 | 74 | |
Set Membership: |
|