Assay Details
| Gene Symbol: gsp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269321.3 | NP_001256250.1 | 5 - 6 | 907 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gsp-1| Gene Symbol: gsp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269321.3 | NP_001256250.1 | 5 - 6 | 907 | 96 | |
Set Membership: |
|