Assay Details
| Gene Symbol: mre-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073335.7 | NP_505736.2 | 8 - 9 | 1848 | 119 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mre-11| Gene Symbol: mre-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073335.7 | NP_505736.2 | 8 - 9 | 1848 | 119 | |
Set Membership: |
|