Assay Details
| Gene Symbol: wdr-5.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029128.4 | NP_001024299.1 | 3 - 4 | 1210 | 162 | |
| NR_002408.1 | 3 - 4 | 1201 | 162 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

wdr-5.3| Gene Symbol: wdr-5.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029128.4 | NP_001024299.1 | 3 - 4 | 1210 | 162 | |
| NR_002408.1 | 3 - 4 | 1201 | 162 | |||
Set Membership: |
|