Assay Details
| Gene Symbol: wdr-5.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029128.4 | NP_001024299.1 | 4 - 5 | 1320 | 146 | |
| NR_002408.1 | 4 - 5 | 1311 | 146 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

wdr-5.3| Gene Symbol: wdr-5.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029128.4 | NP_001024299.1 | 4 - 5 | 1320 | 146 | |
| NR_002408.1 | 4 - 5 | 1311 | 146 | |||
Set Membership: |
|