Assay Details
| Gene Symbol: nrx-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129396.5 | NP_001122868.2 | 21 - 22 | 3951 | 69 | |
| NM_001269333.4 | NP_001256262.1 | 21 - 22 | 3954 | 69 | ||
| NM_001276930.3 | NP_001263859.1 | 22 - 23 | 3968 | 69 | ||
| NM_001276931.3 | NP_001263860.1 | 22 - 23 | 3968 | 69 | ||
More Information
Set Membership: |
|

nrx-1
Genomic Map