Assay Details
| Gene Symbol: nrx-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129396.5 | NP_001122868.2 | 27 - 28 | 5025 | 75 | |
| NM_001269333.4 | NP_001256262.1 | 26 - 27 | 4500 | 75 | ||
| NM_001269334.3 | NP_001256263.1 | 7 - 8 | 1097 | 75 | ||
| NM_001269335.3 | NP_001256264.1 | 6 - 7 | 569 | 75 | ||
| NM_001269336.2 | NP_001256265.1 | 6 - 7 | 1277 | 75 | ||
| NM_001269337.3 | NP_001256266.1 | 6 - 7 | 869 | 75 | ||
| NM_001269341.3 | NP_001256270.1 | 3 - 4 | 278 | 75 | ||
| NM_001276930.3 | NP_001263859.1 | 28 - 29 | 5042 | 75 | ||
| NM_001276931.3 | NP_001263860.1 | 27 - 28 | 4514 | 75 | ||
| NM_001307745.4 | NP_001294674.1 | 5 - 6 | 924 | 75 | ||
| NM_001377602.2 | NP_001364673.1 | 4 - 5 | 539 | 75 | ||
| GenBank mRNA | KF227795.1 | - | 5 - 6 | 728 | 75 | |
More Information
Set Membership: |
|

nrx-1
Genomic Map