Assay Details
| Gene Symbol: F17C11.26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001347268.3 | NP_001334192.1 | 3 - 4 | 277 | 66 | |
| Gene Symbol: zip-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380751.1 | NP_001368624.1 | 3 - 4 | 288 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001380751.1 transcripts.
Set Membership: |
|

F17C11.26
Genomic Map