Assay Details
| Gene Symbol: pak-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073408.6 | NP_505809.2 | 2 - 3 | 278 | 93 | |
| GenBank mRNA | DQ523831.1 | - | 2 - 3 | 300 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pak-2| Gene Symbol: pak-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073408.6 | NP_505809.2 | 2 - 3 | 278 | 93 | |
| GenBank mRNA | DQ523831.1 | - | 2 - 3 | 300 | 93 | |
Set Membership: |
|