Assay Details
| Gene Symbol: pak-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073408.6 | NP_505809.2 | 5 - 6 | 1369 | 136 | |
| NM_073409.6 | NP_505810.2 | 6 - 7 | 1440 | 136 | ||
| GenBank mRNA | DQ523831.1 | - | 5 - 6 | 1391 | 136 | |
More Information
Set Membership: |
|

pak-2
Genomic Map