Assay Details
| Gene Symbol: ima-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383403.1 | 1 - 2 | 94 | 79 | ||
| Gene Symbol: pgap-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073454.6 | NP_505855.2 | 1 - 2 | 66 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_073454.6 transcripts.
Set Membership: |
|

ima-1
Genomic Map