Assay Details
| Gene Symbol: snf-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073472.7 | NP_505873.2 | 2 - 3 | 282 | 66 | |
| GenBank mRNA | AY529124.1 | - | 2 - 3 | 294 | 66 | |
| DQ482732.1 | - | 2 - 3 | 295 | 66 | ||
More Information
Set Membership: |
|

snf-11
Genomic Map