Assay Details
| Gene Symbol: snf-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073472.7 | NP_505873.2 | 4 - 5 | 996 | 78 | |
| GenBank mRNA | AY529124.1 | - | 4 - 5 | 1008 | 78 | |
| DQ482732.1 | - | 4 - 5 | 1009 | 78 | ||
More Information
Set Membership: |
|

snf-11
Genomic Map