Assay Details
| Gene Symbol: grl-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028922.4 | NP_001024093.1 | 1 - 2 | 497 | 60 | |
| NM_001028923.6 | NP_001024094.1 | 1 - 2 | 497 | 60 | ||
More Information
Set Membership: |
|

grl-7
Genomic Map