Assay Details
| Gene Symbol: grl-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028922.4 | NP_001024093.1 | 5 - 6 | 982 | 70 | |
| NM_001028923.6 | NP_001024094.1 | 4 - 5 | 925 | 70 | ||
More Information
Set Membership: |
|

grl-7
Genomic Map