Assay Details
| Gene Symbol: des-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269391.4 | NP_001256320.1 | 8 - 9 | 1222 | 101 | |
| NM_001380763.2 | NP_001368114.1 | 5 - 6 | 880 | 101 | ||
| GenBank mRNA | AF077307.1 | - | 5 - 6 | 895 | 101 | |
More Information
Set Membership: |
|

des-2
Genomic Map