Assay Details
| Gene Symbol: des-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269391.4 | NP_001256320.1 | 9 - 10 | 1369 | 136 | |
| NM_001380763.2 | NP_001368114.1 | 6 - 7 | 1027 | 136 | ||
| GenBank mRNA | AF077307.1 | - | 6 - 7 | 1042 | 136 | |
More Information
Set Membership: |
|

des-2
Genomic Map