Assay Details
| Gene Symbol: des-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269391.4 | NP_001256320.1 | 10 - 11 | 1618 | 59 | |
| NM_001380763.2 | NP_001368114.1 | 7 - 8 | 1276 | 59 | ||
| GenBank mRNA | AF077307.1 | - | 7 - 8 | 1291 | 59 | |
More Information
Set Membership: |
|

des-2
Genomic Map