Assay Details
| Gene Symbol: des-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269391.4 | NP_001256320.1 | 12 - 13 | 2068 | 70 | |
| GenBank mRNA | AF077307.1 | - | 9 - 10 | 1741 | 70 | |
More Information
Set Membership: |
|

des-2
Genomic Map