Assay Details
| Gene Symbol: mig-17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073500.7 | NP_505901.2 | 4 - 5 | 718 | 143 | |
| GenBank mRNA | AB044562.1 | - | 4 - 5 | 699 | 143 | |
More Information
Set Membership: |
|

mig-17
Genomic Map