Assay Details
| Gene Symbol: aat-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129542.4 | NP_001123014.1 | 3 - 4 | 353 | 76 | |
| NM_073504.5 | NP_505905.2 | 4 - 5 | 448 | 76 | ||
More Information
Set Membership: |
|

aat-6
Genomic Map