Assay Details
| Gene Symbol: ttll-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269402.3 | NP_001256331.1 | 3 - 4 | 705 | 129 | |
| NM_001269403.2 | NP_001256332.1 | 2 - 3 | 470 | 129 | ||
More Information
Set Membership: |
|

ttll-5
Genomic Map