Assay Details
| Gene Symbol: ttll-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269402.3 | NP_001256331.1 | 6 - 7 | 1866 | 104 | |
| NM_001269403.2 | NP_001256332.1 | 5 - 6 | 1631 | 104 | ||
More Information
Set Membership: |
|

ttll-5
Genomic Map