Assay Details
| Gene Symbol: nphp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073581.5 | NP_505982.2 | 4 - 5 | 508 | 149 | |
| GenBank mRNA | AY959881.1 | - | 4 - 5 | 405 | 149 | |
More Information
Set Membership: |
|

nphp-4
Genomic Map