Assay Details
| Gene Symbol: nphp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073581.5 | NP_505982.2 | 8 - 9 | 1901 | 154 | |
| GenBank mRNA | AY959881.1 | - | 8 - 9 | 1798 | 154 | |
More Information
Set Membership: |
|

nphp-4
Genomic Map