Assay Details
| Gene Symbol: aman-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073594.4 | NP_505995.2 | 10 - 11 | 3177 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

aman-2| Gene Symbol: aman-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073594.4 | NP_505995.2 | 10 - 11 | 3177 | 70 | |
Set Membership: |
|