Assay Details
| Gene Symbol: snr-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073603.8 | NP_506004.1 | 3 - 4 | 372 | 142 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

snr-4| Gene Symbol: snr-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073603.8 | NP_506004.1 | 3 - 4 | 372 | 142 | |
Set Membership: |
|