Assay Details
| Gene Symbol: add-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047665.4 | NP_001041130.1 | 1 - 2 | 78 | 122 | |
| GenBank mRNA | AF166171.1 | - | 1 - 2 | 78 | 122 | |
More Information
Set Membership: |
|

add-2
Genomic Map