Assay Details
| Gene Symbol: add-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047665.4 | NP_001041130.1 | 7 - 8 | 1459 | 111 | |
| NM_001047666.5 | NP_001041131.1 | 2 - 3 | 508 | 111 | ||
| GenBank mRNA | AF166171.1 | - | 7 - 8 | 1459 | 111 | |
More Information
Set Membership: |
|

add-2
Genomic Map