Assay Details
| Gene Symbol: add-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047665.4 | NP_001041130.1 | 8 - 9 | 1709 | 68 | |
| NM_001047666.5 | NP_001041131.1 | 3 - 4 | 758 | 68 | ||
| GenBank mRNA | AF166171.1 | - | 8 - 9 | 1709 | 68 | |
More Information
Set Membership: |
|

add-2
Genomic Map