Assay Details
| Gene Symbol: atn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269494.3 | NP_001256423.1 | 1 - 2 | 229 | 79 | |
| NM_001269495.3 | NP_001256424.1 | 1 - 2 | 229 | 79 | ||
| NM_073726.6 | NP_506127.1 | 2 - 3 | 445 | 79 | ||
| NM_073727.5 | NP_506128.1 | 2 - 3 | 447 | 79 | ||
More Information
Set Membership: |
|

atn-1
Genomic Map