Assay Details
| Gene Symbol: atn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269494.3 | NP_001256423.1 | 4 - 5 | 739 | 108 | |
| NM_001269495.3 | NP_001256424.1 | 4 - 5 | 661 | 108 | ||
| NM_073726.6 | NP_506127.1 | 5 - 6 | 955 | 108 | ||
| NM_073727.5 | NP_506128.1 | 5 - 6 | 879 | 108 | ||
More Information
Set Membership: |
|

atn-1
Genomic Map