Assay Details
| Gene Symbol: atn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269495.3 | NP_001256424.1 | 2 - 3 | 483 | 128 | |
| NM_073727.5 | NP_506128.1 | 3 - 4 | 701 | 128 | ||
More Information
Set Membership: |
|

atn-1
Genomic Map