Assay Details
| Gene Symbol: cal-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269499.3 | NP_001256428.1 | 2 - 3 | 470 | 106 | |
| NM_001269500.2 | NP_001256429.1 | 1 - 2 | 370 | 106 | ||
More Information
Set Membership: |
|

cal-1
Genomic Map