Assay Details
| Gene Symbol: sel-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073743.7 | NP_506144.1 | 2 - 3 | 238 | 66 | |
| GenBank mRNA | U50829.1 | - | 1 - 2 | 140 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sel-1| Gene Symbol: sel-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073743.7 | NP_506144.1 | 2 - 3 | 238 | 66 | |
| GenBank mRNA | U50829.1 | - | 1 - 2 | 140 | 66 | |
Set Membership: |
|