Assay Details
| Gene Symbol: sel-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073743.7 | NP_506144.1 | 5 - 6 | 1347 | 66 | |
| GenBank mRNA | U50829.1 | - | 4 - 5 | 1249 | 66 | |
| Gene Symbol: mrps-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073744.6 | NP_506145.2 | 1 - 2 | 483 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_073744.6 transcripts.
Set Membership: |
|

sel-1
Genomic Map