Assay Details
| Gene Symbol: ZC443.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073804.4 | NP_506205.1 | 4 - 5 | 945 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZC443.1| Gene Symbol: ZC443.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073804.4 | NP_506205.1 | 4 - 5 | 945 | 73 | |
Set Membership: |
|