Assay Details
| Gene Symbol: anp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047695.5 | NP_001041160.1 | 2 - 3 | 183 | 75 | |
| NM_001047696.3 | NP_001041161.1 | 1 - 2 | 147 | 75 | ||
More Information
Set Membership: |
|

anp-1
Genomic Map