Assay Details
| Gene Symbol: anp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047695.5 | NP_001041160.1 | 6 - 7 | 1114 | 102 | |
| NM_001047696.3 | NP_001041161.1 | 5 - 6 | 1078 | 102 | ||
More Information
Set Membership: |
|

anp-1
Genomic Map