Assay Details
| Gene Symbol: fmi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313142.3 | NP_001300071.1 | 18 - 19 | 5582 | 138 | |
| NM_001313143.3 | NP_001300072.1 | 11 - 12 | 3617 | 138 | ||
| NM_073855.6 | NP_506256.3 | 22 - 23 | 6686 | 138 | ||
| GenBank mRNA | AY314773.1 | - | 22 - 23 | 6679 | 138 | |
More Information
Set Membership: |
|

fmi-1
Genomic Map