Assay Details
| Gene Symbol: fmi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313142.3 | NP_001300071.1 | 1 - 2 | 473 | 122 | |
| NM_073855.6 | NP_506256.3 | 5 - 6 | 1577 | 122 | ||
| GenBank mRNA | AY314773.1 | - | 5 - 6 | 1570 | 122 | |
More Information
Set Membership: |
|

fmi-1
Genomic Map