Assay Details
| Gene Symbol: fmi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313142.3 | NP_001300071.1 | 2 - 3 | 577 | 85 | |
| NM_073855.6 | NP_506256.3 | 6 - 7 | 1681 | 85 | ||
| GenBank mRNA | AY314773.1 | - | 6 - 7 | 1674 | 85 | |
More Information
Set Membership: |
|

fmi-1
Genomic Map