Assay Details
| Gene Symbol: scyl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380793.2 | NP_001366843.1 | 10 - 11 | 2434 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

scyl-1| Gene Symbol: scyl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380793.2 | NP_001366843.1 | 10 - 11 | 2434 | 68 | |
Set Membership: |
|