Assay Details
| Gene Symbol: srr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313070.4 | NP_001299999.1 | 6 | 1070 | 118 | |
| NM_001377607.2 | NP_001364593.1 | 6 - 7 | 1066 | 118 | ||
More Information
Set Membership: |
|

srr-1
Genomic Map